siRNA Details
Gene Symbol: Zfp280c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166648.1 | NP_001160120.1 | 7 | 627 | ||
NM_001166649.1 | NP_001160121.1 | 7 | 648 | |||
NM_001166650.1 | NP_001160122.1 | 6 | 586 | |||
NM_153532.3 | NP_705760.2 | 6 | 958 | |||
GenBank mRNA | AK129399.1 | - | 7 | 607 | ||
AK141491.1 | - | 7 | 628 | |||
BC028839.1 | - | 6 | 598 | 5150255 | ||
BC051397.1 | - | 6 | 586 | 5150255 | ||
BC063775.1 | - | 6 | 970 | 5375514 |
