siRNA Details
Gene Symbol: Shroom2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290684.1 | NP_001277613.1 | 8 | 3829 | ||
NM_001290685.1 | NP_001277614.1 | 4 | 1154 | |||
NM_001290686.1 | NP_001277615.1 | 4 | 961 | |||
NM_001290687.1 | NP_001277616.1 | 4 | 1143 | |||
NM_172441.3 | NP_766029.2 | 8 | 3847 | |||
GenBank mRNA | AK029338.1 | - | 4 | 835 | ||
AK032256.1 | - | 4 | 836 | |||
AK049850.1 | - | 4 | 893 | |||
BC092373.1 | - | 4 | 1149 | |||
EF071946.1 | - | 8 | 3847 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
