siRNA Details
Gene Symbol: Zfp871 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172458.3 | NP_766046.3 | 4 | 386 | ||
GenBank mRNA | AI119099.1 | - | 4 | 386 | ||
AK029041.1 | - | 4 | 374 | |||
AK122579.1 | - | 4 | 349 | |||
BC043671.1 | - | 4 | 501 | 5040597 |
