siRNA Details
Gene Symbol: Frmd4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177843.1 | NP_001171314.1 | 10 | 1225 | ||
NM_172475.3 | NP_766063.3 | 10 | 855 | |||
GenBank mRNA | AK043369.1 | - | 10 | 732 | ||
AK078493.1 | - | 10 | 729 | |||
AK082365.1 | - | 10 | 801 | |||
BC058672.1 | - | 10 | 846 | 6830456 | ||
BC082808.1 | - | 10 | 1232 |
