siRNA Details
Gene Symbol: Tbc1d4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081278.2 | NP_001074747.2 | 2 | 1305 | ||
GenBank mRNA | AK031120.1 | - | 2 | 1149 | ||
AK039337.1 | - | 2 | 1306 | |||
AK044719.1 | - | 2 | 947 | |||
BC064433.1 | - | 2 | 929 | |||
BC080762.1 | - | 2 | 923 | |||
BC117869.1 | - | 2 | 781 | 40095526 | ||
BC117870.1 | - | 2 | 781 | 40095531 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
