siRNA Details
Gene Symbol: DLC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164271.1 | NP_001157743.1 | 5 | 653 | ||
NM_001316668.1 | NP_001303597.1 | 5 | 841 | |||
NM_006094.4 | NP_006085.2 | 5 | 911 | |||
NM_182643.2 | NP_872584.2 | 9 | 2346 | |||
XM_005273374.1 | XP_005273431.1 | 9 | 2227 | |||
XM_017012951.1 | XP_016868440.1 | 9 | 2097 | |||
XM_017012952.1 | XP_016868441.1 | 5 | 644 | |||
GenBank mRNA | AB051510.1 | - | 9 | 2232 | ||
AF026219.1 | - | 5 | 864 | |||
AF035119.1 | - | 5 | 915 | |||
AK025544.1 | - | 5 | 841 | |||
AK289734.1 | - | 5 | 834 | |||
AK291682.1 | - | 9 | 2187 | |||
AK299049.1 | - | 5 | 653 | |||
BC054511.1 | - | 9 | 2101 | 5240161 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
