siRNA Details
Gene Symbol: Dsp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023842.2 | NP_076331.2 | 24 | 8882 | ||
GenBank mRNA | AK077574.1 | - | 1 | 1384 | ||
BC054853.1 | - | 1 | 534 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
