siRNA Details
Gene Symbol: Mex3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175366.3 | NP_780575.2 | 2 | 2203 | ||
GenBank mRNA | AK077009.1 | - | 1 | 810 | ||
AK173330.1 | - | 1 | 3016 | |||
BC031512.1 | - | 1 | 1187 | |||
BC067199.1 | - | 1 | 1168 | |||
BC108401.1 | - | 1 | 1411 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001191626.
Gene Ontology Categories:
