siRNA Details
Gene Symbol: Mex3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175366.3 | NP_780575.2 | 2 | 2857 | ||
GenBank mRNA | AK077009.1 | - | 1 | 1464 | ||
AK088538.1 | - | 1 | 305 | |||
BC067199.1 | - | 1 | 1823 | |||
BC108401.1 | - | 1 | 2067 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
