siRNA Details
Gene Symbol: Rdh13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290409.1 | NP_001277338.1 | 6 | 2103 | ||
NM_001290411.1 | NP_001277340.1 | 7 | 2241 | |||
NM_175372.4 | NP_780581.1 | 7 | 2208 | |||
GenBank mRNA | AK028434.1 | - | 7 | 2202 | ||
AK078817.1 | - | 1 | 691 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
