siRNA Details
Gene Symbol: Slc38a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178746.4 | NP_848861.1 | 15 | 4428 | ||
GenBank mRNA | AK076463.1 | - | 1 | 616 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
