siRNA Details
Gene Symbol: Rnf169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175388.3 | NP_780597.2 | 6 | 6985 | ||
GenBank mRNA | AK076142.1 | - | 1 | 1113 | ||
AK081673.1 | - | 1 | 2048 | |||
AK085104.1 | - | 1 | 2234 | |||
AK173319.1 | - | 5 | 6490 | |||
BC062943.1 | - | 1 | 4329 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
