siRNA Details
Gene Symbol: Tbck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK046449.1 | - | 1 | 2375 | ||
AK081215.1 | - | 1 | 1050 | |||
BB283956.1 | - | 1 | 113 | |||
BB355593.2 | - | 1 | 210 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
