siRNA Details
Gene Symbol: Prr9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175424.3 | NP_780633.1 | 2 | 817 | ||
GenBank mRNA | AK079463.1 | - | 2 | 818 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Prr9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175424.3 | NP_780633.1 | 2 | 817 | ||
GenBank mRNA | AK079463.1 | - | 2 | 818 |