siRNA Details
Gene Symbol: Tmem132c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175432.3 | NP_780641.2 | 7 | 1863 | ||
GenBank mRNA | AK028289.1 | - | 7 | 1882 | ||
AK028541.1 | - | 7 | 1864 | |||
AK035053.1 | - | 5 | 761 | |||
AK157823.1 | - | 7 | 1864 | |||
AK163356.1 | - | 3 | 430 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
