siRNA Details
Gene Symbol: Tmem132c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175432.3 | NP_780641.2 | 9 | 2623 | ||
GenBank mRNA | AK028289.1 | - | 9 | 2642 | ||
AK028541.1 | - | 9 | 2624 | |||
AK035053.1 | - | 7 | 1521 | |||
AK157823.1 | - | 9 | 2624 | |||
AK163356.1 | - | 5 | 1191 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
