siRNA Details
Gene Symbol: Zkscan2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081329.1 | NP_001074798.1 | 7 | 3153 | ||
GenBank mRNA | AK034954.1 | - | 7 | 3155 | ||
BC028272.1 | - | 1 | 879 | |||
BC054371.1 | - | 1 | 868 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
