siRNA Details
Gene Symbol: Heatr5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081179.1 | NP_001074648.1 | 29 | 4823 | ||
GenBank mRNA | AK052391.1 | - | 3 | 642 | ||
AK141783.1 | - | 526 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
