siRNA Details
Gene Symbol: Setd1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040398.2 | NP_001035488.2 | 9, 10 | 3574 | ||
GenBank mRNA | BC035291.1 | - | 1, 2 | 76 | ||
BC038367.1 | - | 3, 4 | 652 | |||
BC040775.1 | - | 1, 2 | 76 | |||
BC041681.1 | - | 1, 2 | 95 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
