siRNA Details
Gene Symbol: Setd1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040398.2 | NP_001035488.2 | 17 | 6488 | ||
GenBank mRNA | AK122435.1 | - | 9 | 2823 | ||
BC023450.1 | - | 3 | 669 | |||
BC035291.1 | - | 9 | 2990 | |||
BC038367.1 | - | 11 | 3566 | |||
BC040775.1 | - | 9 | 2990 | |||
BC041681.1 | - | 9 | 3009 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
