siRNA Details
Gene Symbol: Thnsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177588.2 | NP_808256.1 | 3 | 480 | ||
GenBank mRNA | AK029518.1 | - | 3 | 481 | ||
AK083895.1 | - | 3 | 480 | |||
BC090667.1 | - | 1 | 189 | 30686917 | ||
BC117854.2 | - | 2 | 264 | 40094242 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
