siRNA Details
Gene Symbol: Tmem164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199357.1 | NP_001186286.1 | 7 | 796 | ||
GenBank mRNA | AK089342.1 | - | 9 | 1426 | ||
AK166488.1 | - | 7 | 782 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
