siRNA Details
Gene Symbol: Nrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK051824.1 | - | 6 | 1412 | ||
AK083943.1 | - | 1 | 121 | |||
AK165104.1 | - | 1 | 132 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
