siRNA Details
Gene Symbol: CKM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001824.4 | NP_001815.2 | 3 | 420 | ||
XM_017026240.1 | XP_016881729.1 | 2 | 305 | |||
GenBank mRNA | AK129878.1 | - | 3 | 318 | ||
AK300730.1 | - | 3 | 318 | |||
AK313281.1 | - | 3 | 318 | |||
BC007462.1 | - | 3 | 318 | 3934232 | ||
BT006793.1 | - | 2 | 245 | |||
DA896166.1 | - | 3 | 418 | |||
DC385783.1 | - | 3 | 421 | |||
M14780.1 | - | 3 | 323 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
