siRNA Details
Gene Symbol: Ank2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178655.3 | NP_848770.2 | 6, 7 | 660 | ||
GenBank mRNA | AK032060.1 | - | 6, 7 | 660 | ||
AK036018.1 | - | 26, 27 | 3000 | |||
AK044634.1 | - | 27, 28 | 3270 | |||
AK082352.1 | - | 6, 7 | 647 | |||
BC059251.1 | - | 6, 7 | 611 | 6405358 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
