siRNA Details
Gene Symbol: Ank2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034168.1 | NP_001029340.1 | 10 | 4667 | ||
NM_178655.3 | NP_848770.2 | 25 | 4970 | |||
GenBank mRNA | AK032060.1 | - | 25 | 4968 | ||
AK050264.1 | - | 1 | 1565 | |||
AK083826.1 | - | 10 | 3228 | |||
BC043123.1 | - | 10 | 4667 | |||
BC059251.1 | - | 25 | 4924 | 6405358 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008761478.
Gene Ontology Categories:
