siRNA Details
Gene Symbol: Pirt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178656.3 | NP_848771.1 | 2 | 910 | ||
GenBank mRNA | AK041182.1 | - | 2 | 911 | ||
AK051158.1 | - | 2 | 856 | |||
AK051285.1 | - | 2 | 920 | |||
AK051645.1 | - | 2 | 911 | |||
AK084056.1 | - | 2 | 875 | |||
EU447173.1 | - | 2 | 901 |
