siRNA Details
Gene Symbol: Fam65b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080381.1 | NP_001073850.1 | 15 | 2273 | ||
NM_001286101.1 | NP_001273030.1 | 15 | 2240 | |||
NM_178658.5 | NP_848773.3 | 14 | 2000 | |||
GenBank mRNA | AK031928.1 | - | 15 | 2226 | ||
AK036452.1 | - | 15 | 2220 | |||
AK036695.1 | - | 3 | 890 | |||
AK040887.1 | - | 14 | 1980 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
