siRNA Details
Gene Symbol: Rbms3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172121.1 | NP_001165592.1 | 11 | 1136 | ||
NM_001172122.1 | NP_001165593.1 | 11 | 1659 | |||
NM_001172123.1 | NP_001165594.1 | 11 | 1659 | |||
NM_001172124.1 | NP_001165595.1 | 11 | 1659 | |||
NM_001172126.1 | NP_001165597.1 | 11 | 1659 | |||
NM_178660.4 | NP_848775.2 | 10 | 1608 | |||
GenBank mRNA | AK030478.1 | - | 10 | 1268 | ||
AK030609.1 | - | 11 | 1293 | |||
AK032938.1 | - | 10 | 1605 | |||
AK033005.1 | - | 11 | 1302 | |||
AK041098.1 | - | 11 | 1661 | |||
AK046197.1 | - | 11 | 1137 | |||
AK052196.1 | - | 11 | 1640 | |||
BC033604.1 | - | 6 | 463 | |||
BC108402.1 | - | 10 | 1582 | 6516420 | ||
BC117841.1 | - | 10 | 1048 | 40093225 |
