siRNA Details
Gene Symbol: Prlr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011169.5 | NP_035299.4 | 10 | 5529 | ||
GenBank mRNA | AK087727.1 | - | 1 | 393 | ||
BC096586.1 | - | 1 | 167 | |||
DV053280.1 | - | 1 | 587 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
