siRNA Details
Gene Symbol: NRGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126181.1 | NP_001119653.1 | 4 | 907 | ||
NM_006176.2 | NP_006167.1 | 4 | 904 | |||
GenBank mRNA | BC002835.2 | - | 4 | 795 | 3660038 | |
U89165.1 | - | 4 | 892 | |||
Y09689.1 | - | 4 | 881 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
