siRNA Details
Gene Symbol: C030039L03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112731.1 | NP_001106202.1 | 6 | 979 | ||
NM_198417.2 | NP_940809.2 | 6 | 959 | |||
GenBank mRNA | AK140966.1 | - | 6 | 883 | ||
BC060708.1 | - | 6 | 959 | 6406744 | ||
BQ770435.1 | - | 1 | 411 |
