siRNA Details
Gene Symbol: Trp53rk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023815.4 | NP_076304.2 | 2 | 674 | ||
GenBank mRNA | AB028045.1 | - | 2 | 664 | ||
AK011574.2 | - | 2 | 635 | |||
AK028411.1 | - | 2 | 675 | |||
AK046261.1 | - | 2 | 652 | |||
AK049291.1 | - | 2 | 659 | |||
BC017155.1 | - | 2 | 692 | 4910610 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
