siRNA Details
Gene Symbol: 1500015O10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024283.3 | NP_077245.2 | 4 | 621 | ||
GenBank mRNA | AK018770.1 | - | 4 | 376 | ||
AK049805.1 | - | 4 | 622 | |||
BC002254.1 | - | 4 | 554 | 3493553 |
