siRNA Details
Gene Symbol: 0610010K14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177601.1 | NP_001171072.1 | 6 | 800 | ||
NM_001177603.1 | NP_001171074.1 | 5 | 695 | |||
NM_001177606.1 | NP_001171077.1 | 4 | 593 | |||
NM_001177607.1 | NP_001171078.1 | 4 | 620 | |||
NM_026757.2 | NP_081033.1 | 5 | 694 | |||
NM_145758.1 | NP_665701.1 | 6 | 796 | |||
GenBank mRNA | AA189586.1 | - | 3 | 306 | ||
AK003842.1 | - | 5 | 694 | |||
AK169173.1 | - | 4 | 584 | |||
AK170380.1 | - | 5 | 693 | |||
BC055357.1 | - | 5 | 635 | 5325264 | ||
CB192659.1 | - | 5 | 696 |
