siRNA Details
Gene Symbol: Fam192a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028221.4 | NP_082497.2 | 5 | 587 | ||
GenBank mRNA | AK010053.1 | - | 5 | 552 | ||
AK033988.1 | - | 5 | 518 | |||
AK050704.1 | - | 5 | 522 | |||
AK151603.1 | - | 5 | 541 | |||
BC016088.1 | - | 4 | 327 | 4503452 | ||
BC023158.1 | - | 5 | 426 | 4946201 | ||
BC025472.1 | - | 5 | 563 | 5353148 | ||
BC027060.1 | - | 5 | 549 | 5358022 | ||
BC027818.1 | - | 4 | 355 | 5320967 | ||
BC040818.1 | - | 540 | 4194531 |
