siRNA Details
Gene Symbol: Trappc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164641.1 | NP_001158113.1 | 15 | 2424 | ||
NM_001164642.1 | NP_001158114.1 | 14 | 2397 | |||
NM_029640.2 | NP_083916.1 | 14 | 1652 | |||
NM_180662.2 | NP_850993.2 | 14 | 2397 | |||
GenBank mRNA | AK028504.1 | - | 14 | 2239 | ||
AK029106.1 | - | 15 | 2266 | |||
AK156739.1 | - | 15 | 2222 | |||
AK173287.1 | - | 4 | 396 | |||
AY630620.1 | - | 14 | 1650 | |||
BC034590.1 | - | 14 | 1650 | 5364921 | ||
BC038938.1 | - | 8 | 967 | |||
BC070463.1 | - | 14 | 2397 | 30613261 |
