siRNA Details
Gene Symbol: PRB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC096209.3 | - | 5 | 725 | 40004740 | |
BC096210.3 | - | 5 | 725 | 40004741 | ||
BC096211.1 | - | 4 | 725 | 40004743 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
