siRNA Details
Gene Symbol: Dok5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163686.1 | NP_001157158.1 | 2 | 424 | ||
NM_029761.4 | NP_084037.3 | 2 | 424 | |||
GenBank mRNA | AF418208.1 | - | 2 | 286 | ||
AK012430.1 | - | 2 | 100 | |||
AK044148.1 | - | 2 | 292 | |||
BC066822.1 | - | 2 | 424 | 30021902 | ||
BY248468.1 | - | 2 | 340 |
