siRNA Details
Gene Symbol: Gm6182 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK008984.1 | - | 6 | 795 | ||
AK166086.1 | - | 5 | 698 | |||
BC048160.1 | - | 5 | 661 | |||
BC051922.1 | - | 5 | 661 | |||
BC059210.1 | - | 4 | 1795 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
