siRNA Details
Gene Symbol: 9530003J23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029906.3 | NP_084182.2 | 4 | 532 | ||
GenBank mRNA | AK020538.1 | - | 4 | 458 | ||
AK035441.1 | - | 4 | 506 | |||
BC120771.1 | - | 4 | 453 | 40129694 | ||
BC120797.1 | - | 4 | 453 | 40129720 |
