siRNA Details
Gene Symbol: Vps33a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029929.3 | NP_084205.3 | 7 | 906 | ||
GenBank mRNA | AF439858.1 | - | 7 | 890 | ||
AK019463.1 | - | 7 | 890 | |||
AK041982.1 | - | 7 | 902 | |||
AK140553.1 | - | 7 | 895 | |||
AK150282.1 | - | 7 | 898 | |||
AK150550.1 | - | 7 | 898 | |||
BC046417.1 | - | 7 | 897 | 4501207 |

More Information
Other Species Targeted

Gene Ontology Categories:
