siRNA Details
Gene Symbol: CD2BP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243646.1 | NP_001230575.1 | 2 | 417 | ||
NM_006110.2 | NP_006101.1 | 3 | 340 | |||
GenBank mRNA | AF104222.1 | - | 3 | 284 | ||
AK315708.1 | - | 3 | 225 | |||
BC000495.2 | - | 3 | 239 | 2820690 | ||
BC001947.1 | - | 3 | 239 | 2820690 | ||
BC013854.1 | - | 2 | 417 | |||
CB135964.1 | - | 3 | 342 | |||
KU178518.1 | - | 2 | 164 | |||
KU178519.1 | - | 2 | 164 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
