siRNA Details
Gene Symbol: Cntrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012018.2 | NP_036148.2 | 4 | 612 | ||
GenBank mRNA | AK020148.1 | - | 4 | 596 | ||
AK031338.1 | - | 4 | 595 | |||
AK041715.1 | - | 4 | 613 | |||
BC038386.1 | - | 4 | 590 | |||
BC158010.1 | - | 3 | 485 | 9007318 |

More Information
Other Species Targeted

Gene Ontology Categories:
