siRNA Details
Gene Symbol: Cntrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012018.2 | NP_036148.2 | 4, 5 | 656 | ||
GenBank mRNA | AK020148.1 | - | 4, 5 | 640 | ||
AK031338.1 | - | 4, 5 | 639 | |||
AK041715.1 | - | 4, 5 | 657 | |||
BC038386.1 | - | 4, 5 | 634 | |||
BC158010.1 | - | 3, 4 | 529 | 9007318 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
