siRNA Details
Gene Symbol: Cntrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK041715.1 | - | 11 | 1822 | ||
BC038386.1 | - | 11 | 1799 | |||
BC038550.1 | - | 2 | 280 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
