siRNA Details
Gene Symbol: Nol3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030152.4 | NP_084428.1 | 4 | 1723 | ||
GenBank mRNA | AK053846.1 | - | 4 | 1724 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
