siRNA Details
Gene Symbol: Sfi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030207.2 | NP_084483.2 | 6, 7 | 670 | ||
GenBank mRNA | AK046135.1 | - | 6, 7 | 667 | ||
AK046373.1 | - | 6, 7 | 667 | |||
AK129159.1 | - | 6, 7 | 609 | |||
AK133558.1 | - | 6, 7 | 618 | |||
AK170090.1 | - | 6, 7 | 630 | |||
BC046305.1 | - | 6, 7 | 618 | 6493676 | ||
BC099677.1 | - | 6, 7 | 646 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
