siRNA Details
Gene Symbol: Sfi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030207.2 | NP_084483.2 | 17, 18 | 1925 | ||
GenBank mRNA | AK046135.1 | - | 12, 13 | 1431 | ||
AK046373.1 | - | 12, 13 | 1431 | |||
AK133558.1 | - | 17, 18 | 1873 | |||
AK170090.1 | - | 13, 14 | 1461 | |||
BC026390.1 | - | 7, 8 | 662 | |||
BC046305.1 | - | 11, 12 | 1189 | 6493676 | ||
BC048950.1 | - | 1, 2 | 49 | |||
BC099677.1 | - | 16, 17 | 1770 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
