siRNA Details
Gene Symbol: N4bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030563.2 | NP_085040.2 | 2 | 1993 | ||
GenBank mRNA | AK044775.1 | - | 1 | 342 | ||
AK147522.1 | - | 2 | 1994 | |||
AK172981.1 | - | 2 | 1975 | |||
BC004022.1 | - | 1 | 768 | |||
BC089324.1 | - | 1 | 1158 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
